Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CXCR2/IL-8RB Antibody (866614), PE/Cy7, Novus Biologicals™
SDP

Catalog No. NB310997 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB310997 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB310997 Supplier Novus Biologicals Supplier No. FAB8110PECY7
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CXCR2/IL-8RB Monoclonal specifically detects CXCR2/IL-8RB in Rat samples. It is validated for Flow Cytometry.

Specifications

Antigen CXCR2/IL-8RB
Applications Flow Cytometry
Classification Monoclonal
Clone 866614
Conjugate PE-Cyanine7
Dilution Flow Cytometry
Formulation PBS
Gene Alias CD182 antigen, CDw128b, chemokine (C-X-C motif) receptor 2, chemokine (CXC) receptor 2, C-X-C chemokine receptor type 2, CXC-R2, CXCR-2, CXCR2 gene for IL8 receptor type B, GRO/MGSA receptor, High affinity interleukin-8 receptor B, IL-8 receptor type 2, IL-8R B, IL8R2, IL8RA, interleukin 8 receptor B, interleukin 8 receptor type 2, interleukin-8 receptor type B
Host Species Mouse
Immunogen NS0 mouse myeloma cell line transfected with rat CXCR2/IL-8 RB, Accession # P35407
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Angiogenesis, Cancer, Chemokines and Cytokines, Cytokine Research, GPCR, Immunology, Innate Immunity, Myeloid derived Suppressor Cell
Primary or Secondary Primary
Gene ID (Entrez) 3579
Target Species Rat
Content And Storage Store at 4C in the dark. Do not freeze.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.