Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CXCR2/IL-8RB Antibody (RM0212-24D12), DyLight 488, Novus Biologicals™
SDP

Catalog No. NBP212238G Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP212238G 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP212238G Supplier Novus Biologicals Supplier No. NBP212238G

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Rat Monoclonal Antibody

CXCR2/IL-8RB Monoclonal specifically detects CXCR2/IL-8RB in Mouse samples. It is validated for Flow Cytometry.

Specifications

Antigen CXCR2/IL-8RB
Applications Flow Cytometry
Classification Monoclonal
Clone RM0212-24D12
Conjugate DyLight 488
Dilution Flow Cytometry
Gene Alias CD182 antigen, CDw128b, chemokine (C-X-C motif) receptor 2, chemokine (CXC) receptor 2, C-X-C chemokine receptor type 2, CXC-R2, CXCR-2, CXCR2 gene for IL8 receptor type B, GRO/MGSA receptor, High affinity interleukin-8 receptor B, IL-8 receptor type 2, IL-8R B, IL8R2, IL8RA, interleukin 8 receptor B, interleukin 8 receptor type 2, interleukin-8 receptor type B
Gene Symbols CXCR2
Host Species Rat
Immunogen Mouse recombinant CXCR2/IL-8RB
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Angiogenesis, Cancer, Cytokine Research, GPCR, Immunology, Innate Immunity
Primary or Secondary Primary
Gene ID (Entrez) 3579
Target Species Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.