Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CXCR3 Antibody (220803), mFluor Violet 610 SE, Novus Biologicals™
SDP

Catalog No. NB349730 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB349730 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB349730 Supplier Novus Biologicals Supplier No. FAB1685MFV610
Add to Cart
Add to Cart

Rat Monoclonal Antibody

CXCR3 Monoclonal specifically detects CXCR3 in Mouse samples. It is validated for CyTOF, Flow Cytometry.

Specifications

Antigen CXCR3
Applications CyTOF, Flow Cytometry
Classification Monoclonal
Clone 220803
Conjugate mFluor Violet 610 SE
Dilution Flow Cytometry, CyTOF-ready
Formulation 50mM Sodium Borate
Gene Alias CD183, CD183 antigen, chemokine (C-X-C motif) receptor 3, chemokine (C-X-C) receptor 3, CKR-L2IP-10 receptor, CMKAR3, C-X-C chemokine receptor type 3, CXC-R3, CXCR-3, G protein-coupled receptor 9CD182, GPR9Mig-R, Interferon-inducible protein 10 receptor, IP10 receptor, IP10-R, Mig receptor, MigR
Host Species Rat
Immunogen CHO Chinese hamster ovary cell line transfected with mouse CXCR3, Met1-Leu367, Accession # AAC40163
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Chemokines and Cytokines, Cytokine Research, GPCR, Immunology, Innate Immunity
Primary or Secondary Primary
Gene ID (Entrez) 2833
Target Species Mouse
Content And Storage Store at 4C in the dark.
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.