Learn More
Description
Specifications
Specifications
| Antigen | CXCR3 |
| Applications | Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Proximity Ligation Assay, Western Blot |
| Classification | Polyclonal |
| Concentration | 1.0 mg/ml |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1 - 3 ug/mL, Immunohistochemistry, Immunocytochemistry/Immunofluorescence reported in scientific literature (PMID 31887215), Immunohistochemistry-Paraffin 1:10 - 1:500, Immunohistochemistry-Frozen reported in scientific literature (PMID 19234157), Proximity Ligation Assay reported in scientific literature (PMID 34433033) |
| Formulation | PBS with 0.05% Sodium Azide |
| Gene Accession No. | P49682 |
| Gene Alias | CD183, CD183 antigen, chemokine (C-X-C motif) receptor 3, chemokine (C-X-C) receptor 3, CKR-L2IP-10 receptor, CMKAR3, C-X-C chemokine receptor type 3, CXC-R3, CXCR-3, G protein-coupled receptor 9CD182, GPR9Mig-R, Interferon-inducible protein 10 receptor, IP10 receptor, IP10-R, Mig receptor, MigR |
| Gene Symbols | CXCR3 |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
