Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CXCR3 Antibody (49801) [DyLight 350], Novus Biologicals™
SDP

Catalog No. NB399330 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB399330 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB399330 Supplier Novus Biologicals Supplier No. FAB160D
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CXCR3 Monoclonal antibody specifically detects CXCR3 in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, Neutralization, CyTOF

Specifications

Antigen CXCR3
Applications Flow Cytometry, Immunohistochemistry, Neutralization, CyTOF
Classification Monoclonal
Clone 49801
Conjugate DyLight 350
Dilution Flow Cytometry, Immunohistochemistry, Neutralization, CyTOF-ready
Formulation 50mM Sodium Borate
Gene Alias CD183, CD183 antigen, chemokine (C-X-C motif) receptor 3, chemokine (C-X-C) receptor 3, CKR-L2IP-10 receptor, CMKAR3, C-X-C chemokine receptor type 3, CXC-R3, CXCR-3, G protein-coupled receptor 9CD182, GPR9Mig-R, Interferon-inducible protein 10 receptor, IP10 receptor, IP10-R, Mig receptor, MigR
Host Species Mouse
Immunogen NS0 mouse myeloma cell line transfected with human CXCR3, Met1-Leu368, Accession # P49682
Purification Method Protein A or G purified from ascites
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Chemokines and Cytokines, Cytokine Research, GPCR, Immunology, Innate Immunity
Primary or Secondary Primary
Gene ID (Entrez) 2833
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.