Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CXCR3 Antibody - BSA Free, Novus Biologicals™
SDP

Catalog No. NB396439 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB396439 0.025 mg
NBP241250 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB396439 Supplier Novus Biologicals Supplier No. NBP2412500.025MG
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody has been used in 2 publications

CXCR3 Polyclonal specifically detects CXCR3 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen CXCR3
Applications Western Blot, ELISA, Immunocytochemistry, Immunofluorescence
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1 - 2 μg/mL, ELISA, Immunocytochemistry/Immunofluorescence 5 μg/mL
Formulation PBS with 0.02% Sodium Azide
Gene Alias CD183, CD183 antigen, chemokine (C-X-C motif) receptor 3, chemokine (C-X-C) receptor 3, CKR-L2IP-10 receptor, CMKAR3, C-X-C chemokine receptor type 3, CXC-R3, CXCR-3, G protein-coupled receptor 9CD182, GPR9Mig-R, Interferon-inducible protein 10 receptor, IP10 receptor, IP10-R, Mig receptor, MigR
Gene Symbols CXCR3
Host Species Rabbit
Immunogen Antibody was raised against a 19 amino acid peptide near the carboxy terminus of human CXCR3 . The immunogen is located within the last 50 amino acids of CXCR3. Amino Acid Squence: RGLQRQPSSSRRDSSWSET
Molecular Weight of Antigen 43 kDa
Purification Method Affinity Purified
Quantity 0.025 mg
Regulatory Status RUO
Research Discipline Cytokine Research, GPCR, Immunology, Innate Immunity
Primary or Secondary Primary
Gene ID (Entrez) 2833
Test Specificity At least two isoforms of CXCR3 are known to exist; this antibody will detect both isoforms.
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. Aliquot and store at-20°C long term. Avoid freeze/thaw cycles.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.