Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CXCR4 Antibody (12G5R) [PerCP], Novus Biologicals™
SDP

Catalog No. NB399094 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB399094 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB399094 Supplier Novus Biologicals Supplier No. FAB170RC
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CXCR4 Monoclonal antibody specifically detects CXCR4 in Human samples. It is validated for Flow Cytometry, Neutralization

Specifications

Antigen CXCR4
Applications Flow Cytometry, Neutralization
Classification Monoclonal
Clone 12G5R
Conjugate PerCP
Dilution Flow Cytometry, Neutralization
Formulation PBS
Gene Alias CD184, chemokine (C-X-C motif) receptor 4, chemokine (C-X-C motif), receptor 4 (fusin), C-X-C chemokine receptor type 4, CXC-R4, CXCR-4, D2S201ESDF-1 receptor, FB22, Fusin, HM89NPYRL, HSY3RR, LAP3, LCR1, LESTRCD184 antigen, Leukocyte-derived seven transmembrane domain receptor, leukocyte-derived seven-transmembrane-domain receptor, lipopolysaccharide-associated protein 3, neuropeptide Y receptor Y3, NPY3RWHIM, NPYR, NPYY3R, pCXCR4, seven transmembrane helix receptor, seven-transmembrane-segment receptor, spleen, Stromal cell-derived factor 1 receptor
Host Species Mouse
Immunogen CP-MAC-infected SUP-T1 human T cell lymphoblastic lymphoma line, Accession # P61073
Purification Method Protein A or G purified from cell culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Adaptive Immunity, Cancer, Chemokines and Cytokines, Cytokine Research, GPCR, Growth and Development, Immunology, Innate Immunity, Metastasis, Neuronal Cell Markers, Signal Transduction, Stem Cells
Primary or Secondary Primary
Gene ID (Entrez) 7852
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.