Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CXCR5 Antibody (51505), DyLight 594, Novus Biologicals™
SDP

Catalog No. NB367106 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB367106 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB367106 Supplier Novus Biologicals Supplier No. FAB190M
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CXCR5 Monoclonal specifically detects CXCR5 in Human samples. It is validated for CyTOF, Flow Cytometry, Immunocytochemistry, Immunohistochemistry, Neutralization.

Specifications

Antigen CXCR5
Applications CyTOF, Flow Cytometry, Immunocytochemistry, Immunohistochemistry, Neutralization
Classification Monoclonal
Clone 51505
Conjugate DyLight 594
Dilution Flow Cytometry, Immunohistochemistry, Neutralization, Immunocytochemistry, CyTOF-reported
Formulation 50mM Sodium Borate
Gene Alias BLR1MDR-15, Burkitt lymphoma receptor 1, Burkitt lymphoma receptor 1, GTP binding protein (chemokine (C-X-C motif)receptor 5), Burkitt lymphoma receptor 1, GTP-binding protein, CD185, CD185 antigen, chemokine (C-X-C motif) receptor 5, C-X-C chemokine receptor type 5, CXC-R5, MDR15CXCR-5, MGC117347, Monocyte-derived receptor 15
Host Species Mouse
Immunogen NS0 mouse myeloma cell line transfected with human CXCR5, Met1-Phe372, Accession # P32302
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Adaptive Immunity, Chemokines and Cytokines, Cytokine Research, GPCR, Immunology, Innate Immunity
Primary or Secondary Primary
Gene ID (Entrez) 643
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.