Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CXCR6 Antibody (221002), PE/Cy7, Novus Biologicals™
SDP

Catalog No. NB313749 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB313749 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB313749 Supplier Novus Biologicals Supplier No. FAB2145PECY7
Add to Cart
Add to Cart

Rat Monoclonal Antibody

CXCR6 Monoclonal antibody specifically detects CXCR6 in Mouse samples. It is validated for Flow Cytometry

Specifications

Antigen CXCR6
Applications Flow Cytometry
Classification Monoclonal
Clone 221002
Conjugate PE-Cyanine7
Dilution Flow Cytometry
Formulation PBS
Gene Alias BONZOG-protein coupled receptor bonzo, CD186 antigen, CDw186, chemokine (C-X-C motif) receptor 6, C-X-C chemokine receptor type 6, CXC-R6, CXCR-6, G protein-coupled receptor, STRL33G-protein coupled receptor STRL33, TYMSTRCD186
Host Species Rat
Immunogen CHO Chinese hamster ovary cell line transfected with mouse CXCR6, Met1-Leu351, Accession # Q9EQ16
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Chemokines and Cytokines, Cytokine Research, GPCR, Immunology, Innate Immunity, Virology Bacteria and Parasites
Primary or Secondary Primary
Gene ID (Entrez) 10663
Target Species Mouse
Content And Storage Store at 4C in the dark. Do not freeze.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.