Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CXCR6 Antibody (879112R), Janelia Fluor™ 549, Novus Biologicals™
SDP

Catalog No. NB373437 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB373437 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB373437 Supplier Novus Biologicals Supplier No. FAB8196RI
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CXCR6 Monoclonal specifically detects CXCR6 in Rat samples. It is validated for CyTOF, Flow Cytometry.

Specifications

Antigen CXCR6
Applications CyTOF, Flow Cytometry
Classification Monoclonal
Clone 879112R
Conjugate Janelia Fluor 549
Dilution Flow Cytometry, CyTOF-ready
Formulation 50mM Sodium Borate
Gene Alias BONZOG-protein coupled receptor bonzo, CD186 antigen, CDw186, chemokine (C-X-C motif) receptor 6, C-X-C chemokine receptor type 6, CXC-R6, CXCR-6, G protein-coupled receptor, STRL33G-protein coupled receptor STRL33, TYMSTRCD186
Host Species Mouse
Immunogen NS0 mouse myeloma cell line transfected with rat CXCR6
Purification Method Protein A or G purified from cell culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Chemokines and Cytokines, Cytokine Research, GPCR, Immunology, Innate Immunity, Virology Bacteria and Parasites
Primary or Secondary Primary
Gene ID (Entrez) 10663
Target Species Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.