Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CXCR7/RDC-1 Antibody (358426), DyLight 594, Novus Biologicals™
SDP

Catalog No. NB370595 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB370595 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB370595 Supplier Novus Biologicals Supplier No. FAB4227M
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CXCR7/RDC-1 Monoclonal specifically detects CXCR7/RDC-1 in Human samples. It is validated for CyTOF, Flow Cytometry, Immunohistochemistry.

Specifications

Antigen CXCR7/RDC-1
Applications CyTOF, Flow Cytometry, Immunohistochemistry
Classification Monoclonal
Clone 358426
Conjugate DyLight 594
Dilution Flow Cytometry, Immunohistochemistry, CyTOF-ready
Formulation 50mM Sodium Borate
Gene Alias chemokine (C-X-C motif) receptor 7, Chemokine orphan receptor 1CMKOR1C-X-C chemokine receptor type 7, CXC-R7, CXCR-7, G protein-coupled receptor, GPR159G-protein coupled receptor 159, RDC-1, RDC1G-protein coupled receptor RDC1 homolog
Host Species Mouse
Immunogen NS0 mouse myeloma cell line transfected with human CXCR7/RDC-1, Met1-Lys362 (Gly131Ser), Accession # AAA62370
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Chemokines and Cytokines, GPCR
Primary or Secondary Primary
Gene ID (Entrez) 57007
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.