Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CXCR7/RDC-1 Antibody - BSA Free, Novus Biologicals™
SDP

Catalog No. NBP224779SS Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP224779SS 0.025 mg
NBP224779 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP224779SS Supplier Novus Biologicals Supplier No. NBP224779SS
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody has been used in 5 publications

CXCR7/RDC-1 Polyclonal specifically detects CXCR7/RDC-1 in Human, Mouse, Rat, Sheep samples. It is validated for Western Blot, Simple Western, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Flow Cytometry (Negative).

Specifications

Antigen CXCR7/RDC-1
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Flow Cytometry
Classification Polyclonal
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Western Blot 4 - 6 ug/mL, Simple Western 1:100, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin 5 - 10 ug/mL, Flow Cytometry (Negative)
Formulation PBS with 0.05% Sodium Azide
Gene Accession No. P25106
Gene Alias chemokine (C-X-C motif) receptor 7, Chemokine orphan receptor 1CMKOR1C-X-C chemokine receptor type 7, CXC-R7, CXCR-7, G protein-coupled receptor, GPR159G-protein coupled receptor 159, RDC-1, RDC1G-protein coupled receptor RDC1 homolog
Gene Symbols ACKR3
Host Species Rabbit
Immunogen A synthetic peptide corresponding amino acids 106-129 of human CXCR7/RDC1 was used as the immunogen, GenBank NP_064707.1.
Purification Method Peptide affinity purified
Quantity 0.025 mg
Regulatory Status RUO
Research Discipline Chemokines and Cytokines, GPCR
Primary or Secondary Primary
Gene ID (Entrez) 57007
Test Specificity This antibody is reactive to Chimpanzee, Cow, Horse, Human, Monkey, Mouse, Rat.
Target Species Human, Mouse, Rat, Sheep
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.