Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CXorf40A Antibody, Novus Biologicals™
SDP

Catalog No. NB396288 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
25 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB396288 25 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB396288 Supplier Novus Biologicals Supplier No. NBP24685925UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

CXorf40A Polyclonal antibody specifically detects CXorf40A in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)

Specifications

Antigen CXorf40A
Applications Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Dilution Immunohistochemistry 1:20 - 1:50, Immunohistochemistry-Paraffin 1:20 - 1:50
Formulation PBS (pH 7.2), 40% Glycerol
Gene Accession No. Q8TE69
Gene Alias chromosome X open reading frame 40, chromosome X open reading frame 40A, CXorf40, EOLA1, EOLA1Endothelial-overexpressed lipopolysaccharide-associated factor 1, FLJ52212, protein CXorf40A
Host Species Rabbit
Immunogen This antibody was developed against Recombinant Protein corresponding to amino acids: YAGFVLNGIKTVETRWRPLLSSQRNCTIAVHIAHRDWE
Purification Method Immunogen affinity purified
Quantity 25 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 91966
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.