Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CXorf40A Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$382.00
Specifications
Antigen | CXorf40A |
---|---|
Dilution | Immunohistochemistry 1:20 - 1:50, Immunohistochemistry-Paraffin 1:20 - 1:50 |
Applications | Immunohistochemistry, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
CXorf40A Polyclonal antibody specifically detects CXorf40A in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
CXorf40A | |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Rabbit | |
Human | |
Q8TE69 | |
91966 | |
IgG | |
Immunogen affinity purified |
Immunohistochemistry 1:20 - 1:50, Immunohistochemistry-Paraffin 1:20 - 1:50 | |
Polyclonal | |
Purified | |
RUO | |
PBS (pH 7.2), 40% Glycerol | |
chromosome X open reading frame 40, chromosome X open reading frame 40A, CXorf40, EOLA1, EOLA1Endothelial-overexpressed lipopolysaccharide-associated factor 1, FLJ52212, protein CXorf40A | |
This antibody was developed against Recombinant Protein corresponding to amino acids: YAGFVLNGIKTVETRWRPLLSSQRNCTIAVHIAHRDWE | |
Primary | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title