Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CXXC4 Antibody, Novus Biologicals™
SDP

Catalog No. NBP146112 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP146112 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP146112 Supplier Novus Biologicals Supplier No. NBP146112
Add to Cart
Add to Cart

Goat Polyclonal Antibody

CXXC4 Polyclonal specifically detects CXXC4 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin, Peptide ELISA.

Specifications

Antigen CXXC4
Applications Immunohistochemistry, Immunohistochemistry (Paraffin), ELISA
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Dilution Immunohistochemistry 3 - 5 ug/ml, Immunohistochemistry-Paraffin 3 - 5 ug/ml, Peptide ELISA Detection limit 1:32000
Formulation Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide
Gene Accession No. NP_079488.1
Gene Alias CXXC finger protein 4, CXXC-type zinc finger protein 4, Dvl-binding protein IDAX (inhibition of the Dvl and Axin complex), IDAXCXXC finger 4, Inhibition of the Dvl and axin complex protein, MGC149872
Gene Symbols CXXC4
Host Species Goat
Immunogen Peptide with sequence C-GTSLERTPVPSAEA corresponding to C-Terminus (near) according to NP_079488.1.
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 80319
Test Specificity Human, Expected from sequence similarity: Mouse.
Target Species Human
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.