Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CXXC4 Antibody, PerCP, Novus Biologicals™
SDP

Catalog No. NB358162 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB358162 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB358162 Supplier Novus Biologicals Supplier No. NBP176491PCP
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

CXXC4 Polyclonal antibody specifically detects CXXC4 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)

Specifications

Antigen CXXC4
Applications Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate PerCP
Dilution Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry-Paraffin
Formulation PBS
Gene Alias CXXC finger protein 4, CXXC-type zinc finger protein 4, Dvl-binding protein IDAX (inhibition of the Dvl and Axin complex), IDAXCXXC finger 4, Inhibition of the Dvl and axin complex protein, MGC149872
Host Species Rabbit
Immunogen Antibody was raised against an 18 amino acid synthetic peptide near the amino terminus of human CXXC4. The immunogen is located within amino acids 50 - 100 of CXXC4. Amino Acid Squence: NLPERVGTFSAIPALGG
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 80319
Target Species Human, Mouse, Rat
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.