Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cxxc4 Polyclonal Antibody, Active Motif™

Rabbit Polyclonal Antibody
$159.90 - $633.45
Specifications
| Antigen | Cxxc4 |
|---|---|
| Concentration | Lot-Specific |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-199-3613
![]()
|
Active Motif
61680 |
10 μL |
Each for $159.90
|
|
|||||
50-199-3612
![]()
|
Active Motif
61679 |
100 μL |
Each for $633.45
|
|
|||||
Description
CXXC4 is zinc finger protein that binds directly to the PDZ domain of DVL1, a protein involved in the Wnt signaling pathway, through its C-terminal region. CXXC4 interaction with DVL1 competes with DVL1-AXIN binding, although the affinity of DVL1 for CXXC4 is lower than that for AXIN. In mouse fibroblasts, CXXC4 suppressed Wnt3a induction of beta-catenin and inhibited Wnt3a-dependent TCF4 activation, suggesting that CXXC4 acts as a negative regulator of the Wnt signaling pathway and functions between DVL and beta-catenin. Recent reports suggest that decreased CXXC4 expression is associated with kidney cancer metastasis, cell proliferation, reduced apoptosis in response to cancer drugs, and upregulation of genes involved with proliferation, invasion and cell survival, suggesting that CXXC4 plays a critical role in tumor progression in kidney cancer.Specifications
| Cxxc4 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse | |
| Q6NXI8, Q9H2H0 | |
| 319478, 80319 | |
| This antibody was raised against a peptide within the N-terminal region of mouse Cxxc4. | |
| Primary | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| CXXC4 |
| Lot-Specific | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 30% glycerol and 0.035% sodium azide; pH 7.5 | |
| 9330210J02Rik; C030003J12Rik; CXXC finger 4; CXXC finger protein 4; Cxxc4; CXXC-type zinc finger protein 4; Dvl-binding protein IDAX (inhibition of the Dvl and Axin complex); Idax; inhibition of the Dvl and axin complex protein; inhibitor of the Dvl and Axin complex | |
| CXXC4 | |
| IgG | |
| Antigen affinity chromatography | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title