Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytiva Cy3 NHS Ester Mono-Reactive CyDye

Catalog No. p-8601027 Shop All Cytiva Products
Click to view available options
Quantity:
1 mg
5 mg
10 mg

Catalog No. 45-001-188

Supplier: Cytiva PA13101

Only null left
Add to Cart
Add to Cart

Use for highly accurate labeling of amine groups. Cytiva Cy3 NHS Ester Mono-Reactive CyDye contain derivitized CyDyes that carry only one reactive group on each dye molecule for accurate labeling of amine groups, for example on antibodies, peptides, or oligonucleotides.

CyDye fluors are versatile fluorophores designed for a broad range of applications. CyDye fluors produce high sensitivity and highly reproducible performance at a biologically suitable pH range of 3 to 10. The use of multicolor CyDye flours enables multiplexing without crosstalk for high accuracy.

Cy3 NHS Ester provides targeted and specific labeling of amine residues on antibodies, peptides, or oligonucleotides.

  • Readily dissolves in and produces a stable signal in aqueous buffers (no need for organic solvents)
  • High purity and thorough QC testing generate high levels of chromophore and reactive dye content
  • Narrow excitation and emission bands result in discrete signals from each fluor
  • Demonstrates larger polarization range than conventional fluors
  • Fluorescence lifetime of 2.8 ns
  • High fluorescence intensity
  • Low non-specific binding
TRUSTED_SUSTAINABILITY

Specifications

Format Dried
For Use With (Application) For labeling via Amine groups, CyDyes are suitable for use in applications such as Fluorescence resonance energy transfer (FRET), time resolved-Fluorescence resonance energy transfer (TR-FRET) and Fluorescence polarization (FP)
Quantity 1 mg
Product Type Cy3 NHS Ester
Kit Contents Foil packs of pre-measured dried dye
pH Range 3 to 10
Solubility Soluble in aqueous solutions
Content And Storage 2°C to 8°C
Label or Dye Cy3
Color Red
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.