Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CYB561D1 Antibody, DyLight 350, Novus Biologicals™
SDP

Catalog No. NB054686 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB054686 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB054686 Supplier Novus Biologicals Supplier No. NBP298672UV
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

CYB561D1 Polyclonal specifically detects CYB561D1 in Human samples. It is validated for Western Blot.

Specifications

Antigen CYB561D1
Applications Western Blot
Classification Polyclonal
Conjugate DyLight 350
Dilution Western Blot
Formulation 50mM Sodium Borate
Gene Alias cytochrome b-561 domain containing 1, cytochrome b561 domain-containing protein 1, FLJ39035, FLJ44753, MGC138204
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human CYB561D1 fragment.
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 284613
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.