Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CYB5R1 Antibody (OTI2D10), PerCP, Novus Biologicals™
SDP

Catalog No. NB288543 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB288543 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB288543 Supplier Novus Biologicals Supplier No. NBP271946PCP
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CYB5R1 Monoclonal antibody specifically detects CYB5R1 in Human, Mouse, Rat, Canine samples. It is validated for Western Blot

Specifications

Antigen CYB5R1
Applications Western Blot
Classification Monoclonal
Clone OTI2D10
Conjugate PerCP
Dilution Western Blot
Formulation PBS
Gene Alias b5R.1, B5R1, CYB5BR1, cytochrome b5 reductase 1, EC 1.6.2.2, Humb5R2, NAD(P)H:quinone oxidoreductase type 3 polypeptide A2, NQO3A2polypeptide A2
Host Species Mouse
Immunogen Full length human recombinant protein of human CYB5R1(NP_057327) produced in HEK293T cell.
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline metabolism
Primary or Secondary Primary
Gene ID (Entrez) 51706
Target Species Human, Mouse, Rat, Canine
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.