Learn More
Description
Specifications
Specifications
| Antigen | CYBB/NOX2 |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | NL7 |
| Conjugate | APC |
| Dilution | Flow Cytometry |
| Gene Accession No. | P04839 |
| Gene Alias | CGD, CGD91-phox, Cytochrome b(558) subunit beta, cytochrome b-245 heavy chain, cytochrome b-245, beta polypeptide, Cytochrome b558 subunit beta, EC 1.6.3, GP91-1, GP91PHOX, GP91-PHOX, Heme-binding membrane glycoprotein gp91phox, NADPH oxidase 2, Neutrophil cytochrome b 91 kDa polypeptide, NOX2chronic granulomatous disease, p22 phagocyte B-cytochrome, p91-PHOX, Superoxide-generating NADPH oxidase heavy chain subunit |
| Gene Symbols | CYBB |
| Host Species | Mouse |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
