Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CYBB/NOX2 Antibody (NL7), PerCP, Novus Biologicals™
SDP

Catalog No. NBP141012P Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP141012P 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP141012P Supplier Novus Biologicals Supplier No. NBP141012PCP
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CYBB/NOX2 Monoclonal specifically detects CYBB/NOX2 in Human samples. It is validated for Flow Cytometry, Flow (Intracellular).

Specifications

Antigen CYBB/NOX2
Applications Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone NL7
Conjugate PerCP
Dilution Flow Cytometry, Flow (Intracellular)
Gene Accession No. P04839
Gene Alias CGD, CGD91-phox, Cytochrome b(558) subunit beta, cytochrome b-245 heavy chain, cytochrome b-245, beta polypeptide, Cytochrome b558 subunit beta, EC 1.6.3, GP91-1, GP91PHOX, GP91-PHOX, Heme-binding membrane glycoprotein gp91phox, NADPH oxidase 2, Neutrophil cytochrome b 91 kDa polypeptide, NOX2chronic granulomatous disease, p22 phagocyte B-cytochrome, p91-PHOX, Superoxide-generating NADPH oxidase heavy chain subunit
Gene Symbols CYBB
Host Species Mouse
Immunogen Cytochrome B245 solubilised in octyl-beta-glucopyranoside
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1536
Test Specificity This antibody recognises the heavy chain of cytochrome B245. It recognises the native as well as the denatured form of the protein
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.