Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cyclic AMP Antibody, Novus Biologicals™
SDP

Catalog No. NBP11922505 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.05 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP11922505 0.05 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP11922505 Supplier Novus Biologicals Supplier No. NBP1192250.05ML
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Cyclic AMP Polyclonal specifically detects Cyclic AMP in All Species samples. It is validated for ELISA.

Specifications

Antigen Cyclic AMP
Applications ELISA
Classification Polyclonal
Conjugate Unconjugated
Dilution ELISA 1:100-1:2000
Formulation No buffer with Sodium Azide
Gene Alias 3' 5' cyclic adenosine monophosphate, Cyclic adenosine monophosphate, Cyclic AMP
Host Species Rabbit
Immunogen cAMP conjugated to a protein carrier molecule
Purification Method Affinity Purified
Quantity 0.05 mL
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity cyclic AMP
Target Species All species
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.