Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cyclin A1 Antibody (E43.2) - BSA Free, Novus Biologicals™
SDP

Catalog No. NB1002659 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB1002659 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB1002659 Supplier Novus Biologicals Supplier No. NB1002659
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Cyclin A2 Monoclonal specifically detects Cyclin A2 in Human samples. It is validated for Western Blot.

Specifications

Antigen Cyclin A2
Applications Western Blot
Classification Monoclonal
Clone E43.2
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:100 - 1:2000
Gene Alias CCN1CCNAcyclin-A, cyclin A2, Cyclin-A, cyclin-A2
Gene Symbols CCNA2
Host Species Mouse
Immunogen Human Cyclin A1
Purification Method Protein A purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Cancer, Cell Biology, Cell Cycle and Replication, Core ESC Like Genes, Stem Cell Markers, Tumor Suppressors
Primary or Secondary Primary
Gene ID (Entrez) 890
Test Specificity Cyclin A (E43.2).
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2a
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.