Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cyclin A1 Antibody (E70.1), DyLight 488, Novus Biologicals™
SDP

Catalog No. NB1002660G Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB1002660G 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB1002660G Supplier Novus Biologicals Supplier No. NB1002660G
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Cyclin A2 Monoclonal specifically detects Cyclin A2 in Bovine samples. It is validated for Western Blot.

Specifications

Antigen Cyclin A2
Applications Western Blot
Classification Monoclonal
Clone E70.1
Conjugate DyLight 488
Dilution Western Blot
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias CCN1CCNAcyclin-A, cyclin A2, Cyclin-A, cyclin-A2
Gene Symbols CCNA2
Host Species Mouse
Immunogen Bovine Cyclin A1
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Cell Biology, Cell Cycle and Replication, Cellular Markers, Core ESC Like Genes, Stem Cell Markers, Tumor Suppressors
Primary or Secondary Primary
Gene ID (Entrez) 890
Target Species Bovine
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.