Learn More
Description
Specifications
Specifications
| Antigen | Cyclin A1 |
| Applications | Western Blot |
| Classification | Monoclonal |
| Clone | XLA1-1 |
| Conjugate | Alexa Fluor 532 |
| Dilution | Western Blot |
| Formulation | 50 mM sodium borate with 0.05% sodium azide |
| Gene Alias | cyclin A1, cyclin-A1 |
| Gene Symbols | CCNA1 |
| Host Species | Mouse |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
