Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cyclin A1 Antibody (XLA1-3), Alexa Fluor™ 405, Novus Biologicals™
SDP

Catalog No. NB384580 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB384580 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB384580 Supplier Novus Biologicals Supplier No. NBP308766AF405
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Cyclin A1 Monoclonal antibody specifically detects Cyclin A1 in Xenopus samples. It is validated for Western Blot

Specifications

Antigen Cyclin A1
Applications Western Blot
Classification Monoclonal
Clone XLA1-3
Conjugate Alexa Fluor 405
Formulation 50mM Sodium Borate
Gene Alias cyclin A1, cyclin-A1
Host Species Mouse
Immunogen Recombinant full-length Xenopus laevis Cyclin A1 protein (Uniprot: P18606)
Purification Method Protein A or G purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Cancer, Cell Biology, Cell Cycle and Replication, Cellular Markers, Core ESC Like Genes, Stem Cell Markers, Tumor Suppressors
Primary or Secondary Primary
Gene ID (Entrez) 8900
Target Species Xenopus species
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.