Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cyclin A1 Antibody (XLA1-3), DyLight 350, Novus Biologicals™
SDP

Catalog No. NBP250188UV Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP250188UV 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP250188UV Supplier Novus Biologicals Supplier No. NBP250188UV
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Cyclin A1 Monoclonal specifically detects Cyclin A1 in Xenopus samples. It is validated for Western Blot.

Specifications

Antigen Cyclin A1
Applications Western Blot
Classification Monoclonal
Clone XLA1-3
Conjugate DyLight 350
Dilution Western Blot
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias cyclin A1, cyclin-A1
Gene Symbols CCNA1
Host Species Mouse
Immunogen Xenopus Cyclin A1
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Cycle and Replication, Core ESC Like Genes, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 8900
Target Species Xenopus species
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.