Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cyclin A1/Cyclin A2 Rabbit anti-Human, Clone: S07-3D4, Novus Biologicals™
SDP

Catalog No. NB159208 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB159208 100 μg
NB159209 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. NB159208 Supplier Novus Biologicals Supplier No. NBP314867100UL
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Monoclonal Antibody

Cyclin A1/Cyclin A2 Monoclonal antibody specifically detects Cyclin A1/Cyclin A2 in Human samples. It is validated for Western Blot

Specifications

Antigen Cyclin A1/Cyclin A2
Applications Western Blot
Classification Monoclonal
Clone S07-3D4
Conjugate Unconjugated
Dilution Western Blot
Formulation 50nM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA
Host Species Rabbit
Immunogen A synthesized peptide derived from human Cyclin A1/Cyclin A2 (Uniprot#: P20248 P78396)
Purification Method Affinity purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Product Type Antibody
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.