Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cyclin A2 Antibody (CCNA2/2333), DyLight 405, Novus Biologicals™
SDP

Catalog No. NB390642 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB390642 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB390642 Supplier Novus Biologicals Supplier No. NBP308789V
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Cyclin A2 Monoclonal antibody specifically detects Cyclin A2 in Human samples. It is validated for Immunohistochemistry (Paraffin)

Specifications

Antigen Cyclin A2
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone CCNA2/2333
Conjugate DyLight 405
Formulation 50mM Sodium Borate
Gene Alias CCN1CCNAcyclin-A, cyclin A2, Cyclin-A, cyclin-A2
Host Species Mouse
Immunogen Full length recombinant Cyclin A2 protein (Uniprot: P20248)
Purification Method Protein A or G purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Cancer, Cell Biology, Cell Cycle and Replication, Cellular Markers, Core ESC Like Genes, Stem Cell Markers, Tumor Suppressors
Primary or Secondary Primary
Gene ID (Entrez) 890
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.