Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cyclin B1 Antibody (2061A), Alexa Fluor 647, Novus Biologicals™
SDP

Catalog No. MAB6000AFD Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB6000AFD 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAB6000AFD Supplier Novus Biologicals Supplier No. MAB6000AF647
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

Cyclin B1 Monoclonal specifically detects Cyclin B1 in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, Western Blot.

Specifications

Antigen Cyclin B1
Applications Flow Cytometry, Immunohistochemistry, Western Blot
Classification Monoclonal
Clone 2061A
Conjugate Alexa Fluor 647
Dilution Western Blot, Flow Cytometry, Immunohistochemistry
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias CCNB, cyclin B1, G2/mitotic-specific cyclin B1, G2/mitotic-specific cyclin-B1
Gene Symbols CCNB1
Host Species Rabbit
Immunogen Partial recombinant human Cyclin-B1 protein (amino acids 1-91) [UniProt P14635]
Purification Method Protein A or G purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Cancer, Cell Biology, Cell Cycle and Replication, Mitotic Regulators, Tumor Suppressors
Primary or Secondary Primary
Gene ID (Entrez) 891
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.