Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cyclin B1 Antibody (2061A) - Azide and BSA Free, Novus Biologicals™
SDP

Catalog No. MAB6000 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB6000 100 μg
MAB6000SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MAB6000 Supplier Novus Biologicals Supplier No. MAB6000
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

Cyclin B1 Monoclonal specifically detects Cyclin B1 in Human samples. It is validated for CyTOF, Flow Cytometry, ICC/IF (Negative), Immunohistochemistry, Western Blot.

Specifications

Antigen Cyclin B1
Applications CyTOF, Flow Cytometry, ICC/IF (Negative), Immunohistochemistry, Western Blot
Classification Monoclonal
Clone 2061A
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Western Blot 0.05 - 0.1 ug/ml, Flow Cytometry, Immunohistochemistry, ICC/IF (Negative), CyTOF-ready
Formulation PBS with No Preservative
Gene Alias CCNB, cyclin B1, G2/mitotic-specific cyclin B1, G2/mitotic-specific cyclin-B1
Gene Symbols CCNB1
Host Species Rabbit
Immunogen Partial recombinant human Cyclin-B1 protein (amino acids 1-91) [UniProt P14635]
Purification Method Protein A or G purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Cancer, Cell Biology, Cell Cycle and Replication, Mitotic Regulators, Tumor Suppressors
Primary or Secondary Primary
Gene ID (Entrez) 891
Target Species Human
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.