Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cyclin B1 Antibody (2061D), DyLight 350, Novus Biologicals™
SDP

Catalog No. NB377138 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB377138 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB377138 Supplier Novus Biologicals Supplier No. FAB60001D
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

Cyclin B1 Monoclonal specifically detects Cyclin B1 in Human samples. It is validated for Flow Cytometry (Intracellular Staining), Immunohistochemistry, Western Blot.

Specifications

Antigen Cyclin B1
Applications Flow Cytometry (Intracellular Staining), Immunohistochemistry, Western Blot
Classification Monoclonal
Clone 2061D
Conjugate DyLight 350
Dilution Western Blot, Simple Western, Immunohistochemistry, Intracellular Staining by Flow Cytometry
Formulation 50mM Sodium Borate
Gene Alias CCNB, cyclin B1, G2/mitotic-specific cyclin B1, G2/mitotic-specific cyclin-B1
Host Species Rabbit
Immunogen E. coli-derived recombinant human Cyclin B1, Met1-Pro91, Accession # P14635
Purification Method Protein A or G purified from cell culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Adaptive Immunity, B Cell Development and Differentiation Markers, Cancer, Cell Biology, Cell Cycle and Replication, Cytokine Research, Immunology, Mesenchymal Stem Cell Markers, Mitotic Regulators, Myeloid derived Suppressor Cell, Signal Transduction, Stem Cell Lines, Stem Cell Markers, Tumor Suppressors
Primary or Secondary Primary
Gene ID (Entrez) 891
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.