Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cyclin B1 Antibody (V92.1), PE, Novus Biologicals™
SDP

Catalog No. NB211281 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB211281 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB211281 Supplier Novus Biologicals Supplier No. NB1002648PE
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Cyclin B1 Monoclonal antibody specifically detects Cyclin B1 in Human, Mouse, Hamster samples. It is validated for Flow Cytometry

Specifications

Antigen Cyclin B1
Applications Flow Cytometry
Classification Monoclonal
Clone V92.1
Conjugate PE
Formulation PBS
Gene Alias CCNB, cyclin B1, G2/mitotic-specific cyclin B1, G2/mitotic-specific cyclin-B1
Host Species Mouse
Immunogen Hamster Cyclin B1
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Adaptive Immunity, B Cell Development and Differentiation Markers, Cancer, Cell Biology, Cell Cycle and Replication, Cytokine Research, Immunology, Mesenchymal Stem Cell Markers, Mitotic Regulators, Myeloid derived Suppressor Cell, Signal Transduction, Stem Cell Lines, Stem Cell Markers, Tumor Suppressors
Primary or Secondary Primary
Gene ID (Entrez) 891
Target Species Human, Mouse, Hamster
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.