Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cyclin B2 Antibody (X121.10), DyLight 405, Novus Biologicals™
SDP

Catalog No. NB1002612V Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB1002612V 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB1002612V Supplier Novus Biologicals Supplier No. NB1002612V
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Cyclin B2 Monoclonal specifically detects Cyclin B2 in Xenopus species samples. It is validated for Immunohistochemistry, Immunoprecipitation, Western Blot.

Specifications

Antigen Cyclin B2
Applications Immunohistochemistry, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone X121.10
Conjugate DyLight 405
Dilution Western Blot, Immunohistochemistry
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias cyclin B2, G2/mitotic-specific cyclin-B2, HsT17299
Gene Symbols CCNB2
Host Species Mouse
Immunogen Xenopus laevis Cyclin B2
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Cell Cycle and Replication, Core ESC Like Genes, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 9133
Target Species Xenopus species
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.