Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cyclin B2 Antibody (X29.2), DyLight 350, Novus Biologicals™
SDP

Catalog No. NB384112 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB384112 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB384112 Supplier Novus Biologicals Supplier No. NBP308843UV
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Cyclin B2 Monoclonal antibody specifically detects Cyclin B2 in Human, Mouse, Rat, Xenopus samples. It is validated for ELISA

Specifications

Antigen Cyclin B2
Applications ELISA
Classification Monoclonal
Clone X29.2
Conjugate DyLight 350
Formulation 50mM Sodium Borate
Gene Alias cyclin B2, G2/mitotic-specific cyclin-B2, HsT17299
Host Species Mouse
Immunogen Recombinant full-length Xenopus laevis Cyclin B2 protein (Uniprot: O95067)
Purification Method Protein A or G purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Cancer, Cell Cycle and Replication, Core ESC Like Genes, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 9133
Target Species Human, Mouse, Rat, Xenopus species
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.