Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cyclin D1 Antibody (CCND1/3370R), Alexa Fluor™ 594, Novus Biologicals™
SDP

Catalog No. NB389080 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB389080 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB389080 Supplier Novus Biologicals Supplier No. NBP308955AF594
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

Cyclin D1 Monoclonal antibody specifically detects Cyclin D1 in Human samples. It is validated for Immunohistochemistry (Paraffin)

Specifications

Antigen Cyclin D1
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone CCND1/3370R
Conjugate Alexa Fluor 594
Formulation 50mM Sodium Borate
Gene Alias B-cell lymphoma 1 protein, BCL-1, BCL-1 oncogene, BCL1D11S287E, cyclin D1, cyclin D1 (PRAD1: parathyroid adenomatosis 1), G1/S-specific cyclin D1, G1/S-specific cyclin-D1, PRAD1 oncogene, PRAD1B-cell CLL/lymphoma 1, U21B31
Host Species Rabbit
Immunogen A synthetic peptide corresponding to residues within aa 270-295 of Cyclin D1 (Uniprot: P24385 )
Purification Method Protein A or G purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Cancer, Cell Cycle and Replication, Core ESC Like Genes, mTOR Pathway, Stem Cell Markers, Wnt Signaling Pathway
Primary or Secondary Primary
Gene ID (Entrez) 595
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.