Learn More
Description
Specifications
Specifications
| Antigen | Cyclin D1 |
| Applications | Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), Immunoprecipitation |
| Classification | Monoclonal |
| Clone | SPM587 |
| Conjugate | HRP |
| Dilution | Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen |
| Gene Alias | B-cell lymphoma 1 protein, BCL-1, BCL-1 oncogene, BCL1D11S287E, cyclin D1, cyclin D1 (PRAD1: parathyroid adenomatosis 1), G1/S-specific cyclin D1, G1/S-specific cyclin-D1, PRAD1 oncogene, PRAD1B-cell CLL/lymphoma 1, U21B31 |
| Gene Symbols | CCND1 |
| Host Species | Mouse |
| Immunogen | Human recombinant full length cyclin D1 protein (Uniprot: P24385) |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
