Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cyclin D2 Antibody (CCND2/2620), Alexa Fluor™ 488, Novus Biologicals™
SDP

Catalog No. NB390522 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB390522 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB390522 Supplier Novus Biologicals Supplier No. NBP308804AF488
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Cyclin D2 Monoclonal antibody specifically detects Cyclin D2 in Human samples. It is validated for ELISA, Protein Array

Specifications

Antigen Cyclin D2
Applications ELISA, Protein Array
Classification Monoclonal
Clone CCND2/2620
Conjugate Alexa Fluor 488
Formulation 50mM Sodium Borate
Gene Alias cyclin D2, G1/S-specific cyclin D2, G1/S-specific cyclin-D2, KIAK0002, MGC102758
Host Species Mouse
Immunogen Recombinant full-length human Cyclin D2 (CCND2) protein (Uniprot: IDP30279)
Purification Method Protein A or G purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Cell Cycle and Replication, Core ESC Like Genes, Ovarian Carcinoma Cell Markers, Stem Cell Markers, Wnt Signaling Pathway
Primary or Secondary Primary
Gene ID (Entrez) 894
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.