Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cyclin D2 Antibody (OTI5A6), PerCP, Novus Biologicals™
SDP

Catalog No. NB289660 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB289660 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB289660 Supplier Novus Biologicals Supplier No. NBP270344PCP
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Cyclin D2 Monoclonal antibody specifically detects Cyclin D2 in Human, Mouse, Rat samples. It is validated for Western Blot

Specifications

Antigen Cyclin D2
Applications Western Blot
Classification Monoclonal
Clone OTI5A6
Conjugate PerCP
Dilution Western Blot
Formulation PBS
Gene Alias cyclin D2, G1/S-specific cyclin D2, G1/S-specific cyclin-D2, KIAK0002, MGC102758
Host Species Mouse
Immunogen Full length human recombinant protein of human CCND2 (NP_001750) produced in E.coli.
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Cycle and Replication, Core ESC Like Genes, Ovarian Carcinoma Cell Markers, Stem Cell Markers, Wnt Signaling Pathway
Primary or Secondary Primary
Gene ID (Entrez) 894
Target Species Human, Mouse, Rat
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.