Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cyclin D3 Antibody, DyLight 488, Novus Biologicals™
SDP

Catalog No. NB056358 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB056358 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB056358 Supplier Novus Biologicals Supplier No. NBP298894G
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Cyclin D3 Polyclonal specifically detects Cyclin D3 in Human samples. It is validated for Western Blot.

Specifications

Antigen Cyclin D3
Applications Western Blot
Classification Polyclonal
Conjugate DyLight 488
Dilution Western Blot
Formulation 50mM Sodium Borate
Gene Alias cyclin D3, D3-type cyclin, G1/S-specific cyclin D3, G1/S-specific cyclin-D3
Host Species Rabbit
Immunogen Produced in rabbits immunized with a synthetic peptide corresponding to the C-terminus of human Cyclin D3.
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Cell Cycle and Replication, Wnt Signaling Pathway
Primary or Secondary Primary
Gene ID (Entrez) 896
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.