Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cyclin D3 Mouse anti-Human,Canine,Rat, Unconjugated, Clone: DCS-22, MilliporeSigma™
Mouse Monoclonal Antibody
$182.31 - $457.74
Specifications
Antigen | Cyclin D3 |
---|---|
Clone | DCS-22 |
Applications | Dot Blot, ELISA, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot |
Classification | Monoclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MABE19172MI
|
MilliporeSigma
MABE191725UL |
25 μL |
Each for $182.31
|
|
|||||
MABE19171MI
|
MilliporeSigma
MABE1917100UL |
100 μL |
Each for $457.74
|
|
|||||
Description
Cyclin D3 Monoclonal antibody specifically detects Cyclin D3 in Human,Canine,Rat samples. It is validated for Dot Blot, Enzyme Immunoassay (ELISA), Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blotting.Specifications
Cyclin D3 | |
Dot Blot, ELISA, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
Unconjugated | |
Mouse | |
Human, Canine, Rat | |
G1/S-specific cyclin-D3 | |
His-tagged, full-length recombinant human Cyclin D3. | |
Primary | |
Stable for 1 year at 2 to 8°C from date of receipt. | |
∼33 kDa observed; 32.52 kDa calculated. Uncharacterized bands may be observed in some lysate(s). |
DCS-22 | |
Monoclonal | |
Purified | |
Epigenetics and Nuclear Function | |
NP_001751.1 | |
CCND3 | |
IgG1 κ | |
Protein G Purified | |
Clone DCS-22 is a mouse monoclonal antibody that detects human Cyclin D3. It targets an epitope within 20 amino acids from the C-terminal region. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title