Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cyclin D3 Rabbit anti-Human, Mouse, Rat, Clone: 4C5C8, Novus Biologicals™
SDP

Catalog No. NB167710 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
20 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB167710 20 μg
NB167709 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB167710 Supplier Novus Biologicals Supplier No. NBP31630920UL
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

Cyclin D3 Monoclonal antibody specifically detects Cyclin D3 in Human, Mouse, Rat samples. It is validated for Western Blot

Specifications

Antigen Cyclin D3
Applications Western Blot
Classification Monoclonal
Clone 4C5C8
Conjugate Unconjugated
Formulation PBS, 0.05% BSA, 50% glycerol, pH7.3
Gene Alias cyclin D3, D3-type cyclin, G1/S-specific cyclin D3, G1/S-specific cyclin-D3
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 202-292 of human Cyclin D3 (P30281).
Purification Method Affinity purified
Quantity 20 μg
Regulatory Status RUO
Research Discipline Cancer, Cell Cycle and Replication, Wnt Signaling Pathway
Primary or Secondary Primary
Gene ID (Entrez) 896
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.