Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cyclin E1 Antibody (CCNE1/4935R), DyLight 350, Novus Biologicals™
SDP

Catalog No. NB390453 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB390453 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB390453 Supplier Novus Biologicals Supplier No. NBP308810UV
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

Cyclin E1 Monoclonal antibody specifically detects Cyclin E1 in Human samples. It is validated for Western Blot, Immunohistochemistry (Paraffin)

Specifications

Antigen Cyclin E1
Applications Western Blot, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone CCNE1/4935R
Conjugate DyLight 350
Formulation 50mM Sodium Borate
Gene Alias CCNEcyclin Es, cyclin E1, cyclin Et, G1/S-specific cyclin-E1
Host Species Rabbit
Immunogen Recombinant human Cyclin E1 protein fragment (around aa 10-176) (exact sequence is proprietary) (Uniprot: IDP24864)
Purification Method Protein A or G purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Cancer, Cell Cycle and Replication, Cellular Markers, Ovarian Carcinoma Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 898
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.