Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cyclin E1 Rabbit anti-Human, Mouse, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
$518.60
Specifications
Antigen | Cyclin E1 |
---|---|
Concentration | 0.31 mg/mL |
Applications | Flow Cytometry, Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-172-7261
![]() ![]() |
Proteintech Group Inc
115541AP150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
Cyclin E is a regulatory subunit of Cdk2 and controls G1 / S transition during the mammalian cell cycle. Multiple isoforms of Cyclin E are expressed only in tumors but not in the normal tissues, suggesting a post transcriptional regulation of Cyclin E. In vitro analyses indicated that these truncated variant isoforms of Cyclin E are able to phosphorylate histone H1. Alterations in Cyclin E protein have been implicated as indicators of worse prognosis in various cancers.Specifications
Cyclin E1 | |
Flow Cytometry, Western Blot | |
Unconjugated | |
Rabbit | |
Human, Mouse | |
P24864, Q61457 | |
12447, 898 | |
Cyclin E1 Fusion Protein Ag2110 | |
Primary | |
-20°C | |
CCNE1 |
0.31 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
CCNE, CCNE1, cyclin E1, G1/S specific cyclin E1 | |
CCNE1 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title