Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cyclin E2 Antibody, mFluor Violet 500 SE, Novus Biologicals™
SDP

Catalog No. NB362187 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB362187 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB362187 Supplier Novus Biologicals Supplier No. NB500130MFV500
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Cyclin E2 Polyclonal antibody specifically detects Cyclin E2 in Human samples. It is validated for Western Blot

Specifications

Antigen Cyclin E2
Applications Western Blot
Classification Polyclonal
Conjugate mFluor Violet 500 SE
Dilution Western Blot
Formulation 50mM Sodium Borate
Gene Alias CYCE2, cyclin E2, G1/S-specific cyclin E2, G1/S-specific cyclin-E2
Host Species Rabbit
Immunogen Partial residue sequence of Cyclin E2 protein.
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Cell Cycle and Replication, Cellular Markers
Primary or Secondary Primary
Gene ID (Entrez) 9134
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.