Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cyclin E2 Antibody, Novus Biologicals™
SDP

Catalog No. NBP177755 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP177755 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP177755 Supplier Novus Biologicals Supplier No. NBP177755
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Cyclin E2 Polyclonal specifically detects Cyclin E2 in Human, Mouse samples. It is validated for Western Blot, ELISA, Immunoprecipitation.

Specifications

Antigen Cyclin E2
Applications Western Blot, ELISA, Immunoprecipitation
Classification Polyclonal
Concentration 1.17 mg/ml
Conjugate Unconjugated
Dilution Western Blot 1:200-1:2000, ELISA 1:20000-1:85000, Immunoprecipitation 1:100
Formulation 0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2 with 0.01% Sodium Azide
Gene Accession No. Q9Z238
Gene Alias CYCE2, cyclin E2, G1/S-specific cyclin E2, G1/S-specific cyclin-E2
Gene Symbols CCNE2
Host Species Rabbit
Immunogen This affinity purified Cyclin E2 Antibody was prepared from whole rabbit serum produced by repeated immunizations with a synthetic peptide corresponding to amino acids at the carboxyl terminus of the Cyclin E2 protein. (Uniprot: Q9Z238)
Molecular Weight of Antigen 46.67 kDa
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9134
Test Specificity This affinity purified antibody is directed against mouse Cyclin E2 protein. A BLAST analysis was used to suggest cross-reactivity with Cyclin E2 protein.
Target Species Human, Mouse
Content And Storage Store at -20°C short term. Aliquot and store at -80°C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.