Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cyclin H Antibody (3G6), Janelia Fluor™ 646, Novus Biologicals™
SDP

Catalog No. NB161372 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB161372 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. NB161372 Supplier Novus Biologicals Supplier No. NBP259440JF646
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

Cyclin H Monoclonal antibody specifically detects Cyclin H in Human samples. It is validated for Western Blot, ELISA

Specifications

Antigen Cyclin H
Applications Western Blot, ELISA
Classification Monoclonal
Clone 3G6
Conjugate Janelia Fluor 646
Formulation 50mM Sodium Borate
Gene Alias CAK, cyclin H, cyclin-dependent kinase-activating kinase complex subunit, cyclin-H, MO15-associated protein, p34CAK complex subunit, p37CDK-activating kinase complex subunit
Host Species Mouse
Immunogen Recombinant human Cyclin H (1-323aa) purified from E.coli
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Cycle and Replication, DNA Repair, Nucleotide Excision Repair
Primary or Secondary Primary
Gene ID (Entrez) 902
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.