Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cyclin M3 Antibody, DyLight 350, Novus Biologicals™
SDP

Catalog No. NB101426 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB101426 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB101426 Supplier Novus Biologicals Supplier No. NBP297251UV
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Cyclin M3 Polyclonal specifically detects Cyclin M3 in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.

Specifications

Antigen Cyclin M3
Applications Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate DyLight 350
Formulation 50 mM Sodium Borate
Gene Alias ACDP3cyclin-M3, ancient conserved domain protein 3, Ancient conserved domain-containing protein 3, cyclin M3, Cyclin-M3, DKFZp434I1016, FLJ20018, metal transporter CNNM3
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human Cyclin M3 fragment.
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 26505
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.