Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Cyclohexyl hexanoate, 99%

Click to view available options
Quantity:
25g
100g
6243-10-3
C12H22O2
198.31
MFCD00046356
DTNOERNOMHQUCN-UHFFFAOYSA-N
hexanoic acid, cyclohexyl ester,cyclohexyl caproate,unii-v3b77j8gnh,v3b77j8gnh,cyclohexyl n-hexanoate,hexanoic acid cyclohexyl ester,acmc-1b676,4-4-methoxyphenyl-1,3-thiazol-4-amine
80388
cyclohexyl hexanoate
CCCCCC(=O)OC1CCCCC1
6243-10-3
C12H22O2
198.31
MFCD00046356
DTNOERNOMHQUCN-UHFFFAOYSA-N
hexanoic acid, cyclohexyl ester,cyclohexyl caproate,unii-v3b77j8gnh,v3b77j8gnh,cyclohexyl n-hexanoate,hexanoic acid cyclohexyl ester,acmc-1b676,4-4-methoxyphenyl-1,3-thiazol-4-amine
80388
cyclohexyl hexanoate
CCCCCC(=O)OC1CCCCC1
Quantity 25g
Formula Weight 198.31
Percent Purity 99%
Chemical Name or Material Cyclohexyl hexanoate